Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003580.2 | NP_001003580.1 | 3 - 4 | 1054 | 63 | |
XM_005162613.4 | XP_005162670.1 | 3 - 4 | 1071 | 63 | ||
XM_005162614.4 | XP_005162671.1 | 4 - 5 | 1168 | 63 | ||
XM_009297352.3 | XP_009295627.1 | 2 - 3 | 989 | 63 | ||
GenBank mRNA | BC077093.1 | - | 3 - 4 | 1058 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |