Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cthrc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003765.2 | NP_001003765.1 | 3 - 4 | 628 | 67 | |
GenBank mRNA | BC079494.1 | - | 3 - 4 | 632 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |