Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004111.2 | NP_001004111.2 | 32 - 33 | 3451 | 119 | |
XM_021468404.1 | XP_021324079.1 | 33 - 34 | 3597 | 119 | ||
GenBank mRNA | AY618702.1 | - | 32 - 33 | 3449 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |