Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007163.2 | NP_001007164.1 | 6 - 7 | 745 | 117 | |
GenBank mRNA | BC108037.1 | - | 6 - 7 | 771 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |