Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1al2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012371.2 | NP_001012371.2 | 5 - 6 | 684 | 130 | |
XM_005161350.4 | XP_005161407.1 | 6 - 7 | 1021 | 130 | ||
XM_021468817.1 | XP_021324492.1 | 7 - 8 | 897 | 130 | ||
GenBank mRNA | AY835381.1 | - | 5 - 6 | 597 | 130 | |
BC093209.1 | - | 5 - 6 | 685 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |