Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013470.1 | NP_001013488.1 | 2 - 3 | 200 | 79 | |
XM_009300121.3 | XP_009298396.1 | 2 - 3 | 269 | 79 | ||
GenBank mRNA | BC091461.1 | - | 2 - 3 | 200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |