Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015062.2 | NP_001015062.1 | 2 - 3 | 842 | 73 | |
GenBank mRNA | BC115210.1 | - | 2 - 3 | 842 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |