Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017842.2 | NP_001017842.1 | 1 - 2 | 186 | 117 | |
GenBank mRNA | BC092772.1 | - | 1 - 2 | 191 | 117 | |
BC165189.1 | - | 1 - 2 | 122 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |