Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017907.2 | NP_001017907.2 | 5 - 6 | 484 | 97 | |
XM_021469326.1 | XP_021325001.1 | 5 - 6 | 409 | 97 | ||
GenBank mRNA | BC092940.1 | - | 5 - 6 | 485 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |