Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020532.1 | NP_001018368.1 | 3 - 4 | 373 | 86 | |
GenBank mRNA | BC078380.1 | - | 3 - 4 | 359 | 86 | |
BC095067.1 | - | 3 - 4 | 373 | 86 | ||
BC152166.1 | - | 3 - 4 | 373 | 86 | ||
BC165441.1 | - | 3 - 4 | 378 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |