Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: six6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020585.1 | NP_001018421.1 | 1 - 2 | 863 | 52 | |
GenBank mRNA | BC095226.1 | - | 1 - 2 | 825 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |