Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020824.1 | NP_001018660.1 | 6 - 7 | 474 | 103 | |
GenBank mRNA | BC095367.1 | - | 6 - 7 | 474 | 103 | |
DQ285661.1 | - | 6 - 7 | 486 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |