Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030136.2 | NP_001025307.1 | 9 - 10 | 1170 | 85 | |
XM_005160502.4 | XP_005160559.1 | 10 - 11 | 1377 | 85 | ||
XM_005160503.4 | XP_005160560.1 | 10 - 11 | 1571 | 85 | ||
XM_017352491.2 | XP_017207980.1 | 10 - 11 | 1571 | 85 | ||
GenBank mRNA | BC163491.1 | - | 9 - 10 | 1203 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |