Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pacsin1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033728.1 | NP_001028900.1 | 9 - 10 | 1334 | 70 | |
XM_005166140.3 | XP_005166197.1 | 9 - 10 | 1428 | 70 | ||
XM_009302728.3 | XP_009301003.1 | 9 - 10 | 1651 | 70 | ||
GenBank mRNA | BC081593.1 | - | 9 - 10 | 1390 | 70 | |
BC097107.1 | - | 9 - 10 | 1334 | 70 | ||
BC165817.1 | - | 8 - 9 | 1262 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |