Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045133.1 | NP_001038598.1 | 5 - 6 | 2078 | 116 | |
XM_005159134.4 | XP_005159191.1 | 6 - 7 | 2603 | 116 | ||
XM_005159135.4 | XP_005159192.1 | 6 - 7 | 2602 | 116 | ||
XM_005159136.4 | XP_005159193.1 | 6 - 7 | 2596 | 116 | ||
GenBank mRNA | BC134846.1 | - | 6 - 7 | 2380 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |