Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otud5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075110.1 | NP_001068578.1 | 5 - 6 | 1237 | 65 | |
GenBank mRNA | BC124533.1 | - | 5 - 6 | 1237 | 65 | |
BC155304.1 | - | 6 - 7 | 1154 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |