Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075112.2 | NP_001068580.1 | 2 - 3 | 403 | 152 | |
GenBank mRNA | BC124285.1 | - | 2 - 3 | 406 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |