Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077163.1 | NP_001070631.1 | 7 - 8 | 1101 | 87 | |
XM_005156643.4 | XP_005156700.1 | 7 - 8 | 1121 | 87 | ||
XM_017358772.2 | XP_017214261.1 | 6 - 7 | 1284 | 87 | ||
GenBank mRNA | BC085630.1 | - | 7 - 8 | 1072 | 87 | |
BC124538.1 | - | 7 - 8 | 1101 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |