Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077385.2 | NP_001070853.1 | 1 - 2 | 153 | 77 | |
GenBank mRNA | BC124692.1 | - | 1 - 2 | 156 | 77 | |
BC165550.1 | - | 1 - 2 | 110 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |