Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbrm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005168762.4 | XP_005168819.1 | 17 - 18 | 2813 | 132 | |
GenBank mRNA | BC098878.1 | - | 6 - 7 | 1159 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |