Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult2st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078168.2 | NP_001071636.2 | 1 - 2 | 175 | 120 | |
XM_017351367.1 | XP_017206856.1 | 2 - 3 | 246 | 120 | ||
GenBank mRNA | BC083293.1 | - | 1 - 2 | 225 | 120 | |
BC115350.1 | - | 1 - 2 | 175 | 120 | ||
DQ640388.1 | - | 1 - 2 | 129 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |