Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult2st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078168.2 | NP_001071636.2 | 4 - 5 | 616 | 95 | |
XM_017351367.1 | XP_017206856.1 | 5 - 6 | 687 | 95 | ||
GenBank mRNA | BC083293.1 | - | 4 - 5 | 665 | 95 | |
BC115350.1 | - | 4 - 5 | 616 | 95 | ||
DQ640388.1 | - | 4 - 5 | 570 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |