Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082910.1 | NP_001076379.1 | 17 - 18 | 2046 | 95 | |
XM_009294060.3 | XP_009292335.1 | 17 - 18 | 2377 | 95 | ||
XM_017352282.2 | XP_017207771.1 | 16 - 17 | 2248 | 95 | ||
XM_017352285.2 | XP_017207774.1 | 9 - 10 | 1153 | 95 | ||
XM_021468144.1 | XP_021323819.1 | 17 - 18 | 2149 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |