Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hadhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089437.1 | NP_001082906.1 | 3 - 4 | 193 | 117 | |
GenBank mRNA | BC096816.1 | - | 3 - 4 | 218 | 117 | |
BC107619.1 | - | 3 - 4 | 216 | 117 | ||
BC115107.1 | - | 3 - 4 | 204 | 117 | ||
BC121756.1 | - | 3 - 4 | 204 | 117 | ||
BC131856.1 | - | 3 - 4 | 193 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |