Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099245.1 | NP_001092715.1 | 1 - 2 | 221 | 93 | |
GenBank mRNA | BC146745.1 | - | 1 - 2 | 221 | 93 | |
HM210101.1 | - | 1 - 2 | 187 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |