Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099447.1 | NP_001092917.1 | 1 - 2 | 367 | 78 | |
XM_005165493.4 | XP_005165550.1 | 1 - 2 | 893 | 78 | ||
GenBank mRNA | BC171554.1 | - | 1 - 2 | 445 | 78 | |
BC171721.1 | - | 1 - 2 | 444 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |