Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099740.2 | NP_001093210.1 | 1 - 2 | 144 | 67 | |
XM_021468031.1 | XP_021323706.1 | 1 - 2 | 162 | 67 | ||
GenBank mRNA | AJ629191.1 | - | 1 - 2 | 117 | 67 | |
AJ697718.1 | - | 1 - 2 | 117 | 67 | ||
BC045465.1 | - | 1 - 2 | 140 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |