Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camsap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100001.3 | NP_001093471.1 | 17 - 18 | 4362 | 72 | |
XM_005168648.4 | XP_005168705.1 | 16 - 17 | 4413 | 72 | ||
XM_021476204.1 | XP_021331879.1 | 17 - 18 | 4446 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |