Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camsap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100001.3 | NP_001093471.1 | 5 - 6 | 774 | 115 | |
XM_021476204.1 | XP_021331879.1 | 5 - 6 | 825 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |