Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camsap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100001.3 | NP_001093471.1 | 6 - 7 | 807 | 75 | |
XM_021476204.1 | XP_021331879.1 | 6 - 7 | 858 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |