Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100040.1 | NP_001093510.1 | 2 - 3 | 313 | 149 | |
XM_021476296.1 | XP_021331971.1 | 2 - 3 | 316 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |