Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100040.1 | NP_001093510.1 | 3 - 4 | 950 | 85 | |
XM_021476296.1 | XP_021331971.1 | 3 - 4 | 953 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |