Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109825.1 | NP_001103295.1 | 7 - 8 | 975 | 89 | |
GenBank mRNA | DQ986317.1 | - | 7 - 8 | 975 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |