Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131005.2 | NP_571080.1 | 4 - 5 | 477 | 64 | |
GenBank mRNA | BC092712.1 | - | 4 - 5 | 477 | 64 | |
X52502.1 | - | 447 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |