Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpc4apa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131569.2 | NP_571644.1 | 10 - 11 | 1289 | 78 | |
GenBank mRNA | AF130459.1 | - | 10 - 11 | 1290 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |