Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131612.2 | NP_571687.1 | 1 - 2 | 341 | 77 | |
XM_017351360.2 | XP_017206849.1 | 1 - 2 | 332 | 77 | ||
GenBank mRNA | AJ245965.1 | - | 264 | 77 | ||
BC162623.1 | - | 1 - 2 | 341 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |