Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jag1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131863.2 | NP_571938.2 | 5 - 6 | 1015 | 83 | |
GenBank mRNA | AF229451.1 | - | 5 - 6 | 977 | 83 | |
AY221106.1 | - | 5 - 6 | 1016 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |