Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153653.1 | NP_705939.1 | 10 - 11 | 1072 | 96 | |
XM_009295279.3 | XP_009293554.1 | 9 - 10 | 850 | 96 | ||
XM_021468749.1 | XP_021324424.1 | 9 - 10 | 1117 | 96 | ||
GenBank mRNA | AF317643.1 | - | 10 - 11 | 1072 | 96 | |
BC042317.1 | - | 10 - 11 | 1232 | 96 | ||
BC066495.1 | - | 10 - 11 | 1061 | 96 | ||
BC164717.1 | - | 10 - 11 | 1115 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |