Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153659.1 | NP_705945.1 | 3 - 4 | 262 | 69 | |
GenBank mRNA | AY029527.1 | - | 3 - 4 | 262 | 69 | |
BC044351.1 | - | 3 - 4 | 244 | 69 | ||
BC066715.1 | - | 3 - 4 | 202 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |