Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173260.1 | NP_775367.1 | 3 - 4 | 219 | 101 | |
XM_005173300.4 | XP_005173357.1 | 3 - 4 | 383 | 101 | ||
GenBank mRNA | AF506232.1 | - | 3 - 4 | 219 | 101 | |
BC071324.1 | - | 3 - 4 | 183 | 101 | ||
BC154771.1 | - | 3 - 4 | 353 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |