Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199669.1 | NP_955963.1 | 4 - 5 | 446 | 79 | |
GenBank mRNA | BC045299.1 | - | 4 - 5 | 446 | 79 | |
BC164883.1 | - | 3 - 4 | 368 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |