Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199669.1 | NP_955963.1 | 6 - 7 | 586 | 68 | |
GenBank mRNA | BC045299.1 | - | 6 - 7 | 586 | 68 | |
BC164883.1 | - | 5 - 6 | 508 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |