Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199730.1 | NP_956024.1 | 2 - 3 | 348 | 123 | |
GenBank mRNA | BC063970.1 | - | 2 - 3 | 348 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |