Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310061.1 | NP_001296990.1 | 4 - 5 | 797 | 65 | |
NM_001310062.1 | NP_001296991.1 | 4 - 5 | 794 | 65 | ||
GenBank mRNA | AY423033.1 | - | 4 - 5 | 564 | 65 | |
BC091886.1 | - | 4 - 5 | 502 | 65 | ||
BC164969.1 | - | 4 - 5 | 380 | 65 | ||
EH442667.1 | - | 4 - 5 | 820 | 65 | ||
EH561355.1 | - | 1 - 2 | 882 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |