Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200314.2 | NP_956608.2 | 5 - 6 | 1093 | 104 | |
GenBank mRNA | BC050500.1 | - | 5 - 6 | 1069 | 104 | |
BC062835.1 | - | 5 - 6 | 1071 | 104 | ||
CU458959.1 | - | 5 - 6 | 1083 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |