Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200422.1 | NP_956716.1 | 2 - 3 | 677 | 79 | |
GenBank mRNA | BC054643.1 | - | 2 - 3 | 677 | 79 |
Gene Symbol: LOC110437745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468193.1 | XP_021323868.1 | 1 - 2 | 596 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |