Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200422.1 | NP_956716.1 | 8 - 9 | 1297 | 62 | |
GenBank mRNA | BC054643.1 | - | 8 - 9 | 1297 | 62 |
Gene Symbol: LOC110437745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468193.1 | XP_021323868.1 | 7 - 8 | 1216 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |