Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200649.1 | NP_956943.1 | 10 - 11 | 1638 | 151 | |
GenBank mRNA | BC057476.1 | - | 10 - 11 | 1638 | 151 | |
BC164569.1 | - | 9 - 10 | 1462 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |