Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1an | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201496.1 | NP_958904.1 | 1 - 2 | 348 | 120 | |
GenBank mRNA | BC044475.1 | - | 1 - 2 | 348 | 120 | |
BC164149.1 | - | 1 - 2 | 182 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |