Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212933.1 | NP_998098.1 | 2 - 3 | 214 | 106 | |
GenBank mRNA | BC067550.1 | - | 2 - 3 | 214 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |